Learn More
Tin selenide, 99.999% (metals basis)
Description
Tin selenide finds application in low- cost photovoltaics devices and memory-switching devices. It acts as a solid-state lubricant. It is also used for solar cells and anodes for lithium-ion batteries. Further, it is a narrow band-gap semiconductor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécification
Spécification
| Melting Point | 861°C |
| Physical Form | Pieces |
| Quantity | 5 g |
| Assay Percent Range | 99.999% (metals basis) |
| Solubility Information | Insoluble in water. Soluble in aqua regia,alkali sulfides,and alkali selenides. |
| Formula Weight | 197.65 |
| Chemical Name or Material | Tin selenide |
RUO – Research Use Only
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.