missing translation for 'onlineSavingsMsg'
Learn More

Tin shot, 2-4mm, Puratronic, 99.9999% (metals basis), Thermo Scientific Chemicals

Product Code. 11341618
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-31-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]

Specifications

Quantity 5 g
Percent Purity 99.9999%
Grade Puratronic™
Odor Odorless
Form Shot
Assay (metals basis)
Chemical Name or Material Tin shot, 3mm (0.1 in.)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'