missing translation for 'onlineSavingsMsg'
Learn More

Tin wire, 0.5mm (0.02in) dia, Puratronic™, 99.9985% (metals basis), Thermo Scientific Chemicals

44.62 € - 327.63 €

Chemical Identifiers

CAS 7440-31-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11396358
View Documents
Thermo Scientific Alfa Aesar
011906.G1
1 m
44.62 €
1m
Please to purchase this item. Need a web account? Register with us today!
11316368
View Documents
Thermo Scientific Alfa Aesar
011906.G5
5 m
73.26 €
5m
Please to purchase this item. Need a web account? Register with us today!
11306368
View Documents
Thermo Scientific Alfa Aesar
011906.G9
25 m
327.63 €
25m
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-31-5
118.71
ATJFFYVFTNAWJD-UHFFFAOYSA-N
5352426
tin
Sn
MFCD00133862
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
CHEBI:32990
[Sn]

Specifications

7440-31-5
MFCD00133862
ATJFFYVFTNAWJD-UHFFFAOYSA-N
tin
5352426
99.9985%
Odorless
Wire
Tin wire, 0.5mm (0.02 in.) dia.
Sn
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
[Sn]
118.71
CHEBI:32990
Puratronic™
≈1.43g/m
(metals basis)
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 23-141-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only