missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin wire, 2.0mm (0.08in) dia, Puratronic™, 99.9985% (metals basis), Thermo Scientific Chemicals
Form: Approx. 22.9g/m
55.10 € - 572.00 €
Chemical Identifiers
CAS | 7440-31-5 |
---|---|
Molecular Formula | Sn |
Molecular Weight (g/mol) | 118.71 |
MDL Number | MFCD00133862 |
InChI Key | ATJFFYVFTNAWJD-UHFFFAOYSA-N |
Synonym | powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german |
PubChem CID | 5352426 |
ChEBI | CHEBI:32990 |
IUPAC Name | tin |
SMILES | [Sn] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11356358
|
Thermo Scientific Alfa Aesar
011904.BU |
25 cm |
55.10 €
25cm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11332008
|
Thermo Scientific Alfa Aesar
011904.BY |
100 cm |
154.00 €
100cm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11342008
|
Thermo Scientific Alfa Aesar
011904.CC |
500 cm |
572.00 €
500cm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-31-5 | |
118.71 | |
ATJFFYVFTNAWJD-UHFFFAOYSA-N | |
5352426 | |
tin |
Sn | |
MFCD00133862 | |
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german | |
CHEBI:32990 | |
[Sn] |
Specifications
Silver to Gray | |
Silver Gray wire | |
Sn | |
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german | |
[Sn] | |
118.71 | |
CHEBI:32990 | |
Wire |
7440-31-5 | |
99.9985% (Metals basis) | |
MFCD00133862 | |
ATJFFYVFTNAWJD-UHFFFAOYSA-N | |
tin | |
5352426 | |
Odorless | |
Tin |
Safety and Handling
missing translation for 'einecsNumber' : 23-141-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only