missing translation for 'onlineSavingsMsg'
Learn More

Tin wire, 3.2mm (0.13in) dia, 99.95% (metals basis), Thermo Scientific Chemicals

Código de producto. 11376887
missing translation for 'orderingAttributeHoverText'
Quantity:
1 m
5 m
Este artículo no se puede devolver. Vea la política de devoluciones
Este artículo no se puede devolver. Vea la política de devoluciones

Form: approx. 58g/m

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 7440-31-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]

Especificaciones

Color Silver to Gray
Appearance Silver Gray wire
Assay Percent Range 99.95% (Metals basis)
Quantity 5 m
Odor Odorless
Form Wire
Chemical Name or Material Tin

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'