Learn More
Tirasemtiv, MedChemExpress
MedChemExpress Tirasemtiv is an activator of the fast skeletal muscle troponin complex.

70.20 € - 585.90 €
Specifications
Chemical Name or Material | Tirasemtiv |
---|---|
Physical Form | Solid |
CAS | 1005491-05-3 |
Color | Gray |
Purity Grade Notes | Research |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16499477
![]() |
Medchem Express
HY-15964 |
5 mg |
70.20 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16437016
![]() |
Medchem Express
HY-15964 |
100 mg |
585.90 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16436938
![]() |
Medchem Express
HY-15964 |
10 mg |
112.05 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16412997
![]() |
Medchem Express
HY-15964 |
50 mg |
363.15 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tirasemtiv is an activator of the fast skeletal muscle troponin complex.Specifications
Tirasemtiv | |
1005491-05-3 | |
Research | |
Neuroscience-Neuromodulation | |
DMSO : 10.91 mg/mL (47.38 mM; Need ultrasonic) | |
230.27 | |
99.83% |
Solid | |
Gray | |
C12H14N4O | |
CK-2017357 | |
O=C1N(C(CC)CC)C2=NC(C#C)=CN=C2N1 | |
230.27 | |
Research |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.