missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tiron, Indicator, Honeywell Fluka™
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Plastic bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Melting Point | 300°C |
Linear Formula | (OH)2C6H2(SO3Na)2 · H2O |
UN Number | NONH for all modes of transport |
Solubility Information | 1350g/L |
Formula Weight | 332.22g/mol |
Packaging | Plastic bottle |
Color | Violet to White |
pH | 5.0 |
Quantity | 10 g |
Physical Form | Solid |
Show More |
Safety and Handling
Product Identifier
- Tiron
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : 4,5-Dihydroxy-1,3-benzenedisulfonic acidhydrate disodium salt
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction