missing translation for 'onlineSavingsMsg'
Learn More

TISAB IV solution, Honeywell Fluka™

For determination of fluoride

90.17 € - 470.00 €

Specifications

Purity Grade Notes General indicators
Packaging Plastic Bottle
Physical Form Liquid, clear
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16675833
View Documents
Honeywell Fluka
34574H-500ML
500 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16685833
View Documents
Honeywell Fluka
34574H-6X500ML
6 x 500 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Specifications

General indicators
Plastic Bottle
Liquid, clear
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS
Documents

Documents

Product Certifications