Learn More
Bel-Art™ ProCulture Tissue Culture Flask Racks
Prevent the domino effect of flasks tipping one over the other
Brand:  Bel-Art™ ProCulture F18970-0001
 View more versions of this product
Product Code. 11722016
  • 89.32 € / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • These polypropylene racks hold 25mL, 75mL, 50 to 75mL, or 250 to 260mL or similar sized tissue culture flasks in a vertical position
  • Rectangular holes in the top and center plates stabilize the flasks preventing tipping
  • Easily organize flasks to improve processing and to avoid errors in collecting used media, media change, washing of cell monolayer, trypsinization and seeding of cells
  • Grips on each end make rack handling easy and allow for safe transport of flasks to the incubator
  • Steam autoclavable at 121°C (250°F)
Specifications
Specifications
Flask Rack
Flasks (12)
50 to 75mL Flat-sided tissue culture flask
23 x 13.7 x 10.7cm (9.1 x 5.4 x 4.2 in.)
4.2 in.
Autoclavable
Polypropylene
121°C
10.7 cm