Learn More
Bel-Art™ ProCulture Tissue Culture Flask Racks
Prevent the domino effect of flasks tipping one over the other
Brand:  Bel-Art™ ProCulture F18970-0002
 View more versions of this product
Product Code. 11732016
  • 83.65 € / Each
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • These polypropylene racks hold 25cc, 75cc, 50 to 75mL, or 250 to 260mL or similar sized tissue culture flasks in a vertical position
  • Rectangular holes in the top and center plates stabilize the flasks preventing tipping
  • Easily organize flasks to improve processing and to avoid errors in collecting used media, media change, washing of cell monolayer, trypsinization and seeding of cells
  • Grips on each end make rack handling easy and allow for safe transport of flasks to the incubator
  • Steam autoclavable at 121°C (250°F)
Order Info
ForUseWithEquipment
Specifications
Specifications
Flask Rack
Autoclavable
Flasks (5)
250 to 260mL Flasks
121°C
29.7 cm
13.2 cm
5 x 1
White
Polypropylene
1 Each
29.7 x 12.5 x 13.2cm
12.5 cm