missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ TissueLoc™ Cassette IV Biopsy Cassettes

For biopsy tissue samples, these cassettes have a back-edge lid hinge with loose-packed lids that are available in 12 different color varieties

284.00 €

Specifications

Closure Included Yes
Quantity 1000/Cs.
Pore Style Small Squares
Closure Type Separate Lid, Back Hinge
PrintMate Compatible No
View More Specs

Products 12
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
16384156
View Documents
Epredia™
B851925BE
Beige
284.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16394156
View Documents
Epredia™
B851925BL
Blue
284.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16304166
View Documents
Epredia™
B851925GN
Green
284.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16314166
View Documents
Epredia™
B851925GY
Gray
284.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16324166
View Documents
Epredia™
B851925LA
Lavender
284.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16334166
View Documents
Epredia™
B851925LG
Light Green
284.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16344166
View Documents
Epredia™
B851925LI
Lilac
284.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16354166
View Documents
Epredia™
B851925OR
Orange
284.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16364166
View Documents
Epredia™
B851925PK
Pink
284.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16374166
View Documents
Epredia™
B851925TN
Tan
284.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16384166
View Documents
Epredia™
B851925WH
White
284.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16394166
View Documents
Epredia™
B851925YW
Yellow
284.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Printing

  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime

Grossing, Processing and Embedding

  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples

Sectioning

  • Precision engineering for solid connection to microtomes

Archiving

  • Cassette surface texture optimizes scannability for archiving
Specifications

Specifications

Yes
Small Squares
No
1000/Cs.
Separate Lid, Back Hinge
TissueLoc Cassette IV with Color-Matched Lids
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.