missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ TissueLoc™ Cassette IV Biopsy Cassettes in Tube Packs

For biopsy tissue samples, these cassettes have a back-edge lid hinge, loose-packed lids, and are compatible with all Epredia PrintMate™ Cassette Printers

283.00 € - 286.00 €

Specifications

Closure Included Yes
Quantity 750/Cs.
Pore Style Small Squares
Closure Type Separate Lid, Back Hinge
PrintMate Compatible Yes
View More Specs

Products 11
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
16364146
View Documents
Epredia™
Beige
286.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16374146
View Documents
Epredia™
Blue
283.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16384146
View Documents
Epredia™
Green
283.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16394146
View Documents
Epredia™
Gray
283.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16304156
View Documents
Epredia™
B851750LA
Lavender
283.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16314156
View Documents
Epredia™
Light Green
283.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16324156
View Documents
Epredia™
B851750LI
Lilac
283.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16334156
View Documents
Epredia™
Orange
283.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16344156
View Documents
Epredia™
B851750PK
Pink
283.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16354156
View Documents
Epredia™
Tan
283.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16374156
View Documents
Epredia™
Yellow
283.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Printing

  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime

Grossing, Processing and Embedding

  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples

Sectioning

  • Precision engineering for solid connection to microtomes

Archiving

  • Cassette surface texture optimizes scannability for archiving
Specifications

Specifications

Yes
Small Squares
Yes
750/Cs.
Separate Lid, Back Hinge
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.