missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ TissueLoc™ Cassette IV Slotted Cassettes in Tube Packs

Slotted Cassettes are ideal for use with routine tissue samples. They are compatible with all Epredia PrintMate™ Cassette Printers, have a back-edge lid hinge, and loose-packed lids that are available in twelve color varieties.

114.00 €

Specifications

Closure Included Yes
Quantity 750/Cs.
Pore Style Slots
Closure Type Separate Lid, Back Hinge
PrintMate Compatible Yes
View More Specs

Products 12
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
16394036
View Documents
Epredia™
B851720BE
Beige
114.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16304046
View Documents
Epredia™
B851720BL
Blue
114.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16314046
View Documents
Epredia™
B851720GN
Green
114.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16324046
View Documents
Epredia™
B851720GY
Gray
114.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16334046
View Documents
Epredia™
B851720LA
Lavender
114.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16344046
View Documents
Epredia™
B851720LG
Light Green
114.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16354046
View Documents
Epredia™
B851720LI
Lilac
114.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16364046
View Documents
Epredia™
B851720OR
Orange
114.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16374046
View Documents
Epredia™
B851720PK
Pink
114.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16384046
View Documents
Epredia™
B851720TN
Tan
114.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16394046
View Documents
Epredia™
B851720WH
White
114.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16304056
View Documents
Epredia™
B851720YW
Yellow
114.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Printing

  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime

Grossing Processing and Embedding

  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples

Sectioning

  • Precision engineering for solid connection to microtomes

Archiving

  • Cassette surface texture optimizes scannability for archiving
Specifications

Specifications

Yes
Slots
Yes
TissueLoc™ Slotted Cassette
750/Cs.
Separate Lid, Back Hinge
For Use With PrintMate™ Printers
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.