missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Scott™ Facial Tissues
GREENER_CHOICE

Provide trusted Scott™ softness and performance in offices and hotel washrooms that have a high-quality image. Scott™ Facial Tissues project an ambience of comfort and care, while promoting user hygiene and personal standards.

Brand:  Kimberly-Clark™ 8837

45.82 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 12628835

  • 122.00 € / Pack of 2100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Flat box packaging designed complements most environments. Hygienic and economic dispensing, FSC certified tissues can be used with Kimberly Clark Professional™ Wall mounted Facial Tissue Dispenser.
  • White
  • 2-ply facial tissues
  • 100% recycled fiber
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tissue
215 mm
186 mm
White
For reliability in locations that demand consistency, quality and value
2-ply
8.5 in.
7.3 in.
Paper
215 x 186 mm
21 Cartons x 100 Sheets (2,100 total)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kimberly-Clark™ Scott™ Facial Tissues > Dimensions (L x W): 215 x 185 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.