missing translation for 'onlineSavingsMsg'
Learn More

Titanium carbide, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11359307
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Used in preparation of cermets and abrasion-resistant surface coating on metal parts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12070-08-5
Molecular Formula CTi
Molecular Weight (g/mol) 59.88
MDL Number MFCD00011268
InChI Key YXIVWSJCLXKLJL-UHFFFAOYSA-N
IUPAC Name methanidylidynetitaniumylium
SMILES [C-]#[Ti+]

Specifications

Boiling Point 4820°C
Melting Point ∽3140°C
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.5% (metals basis)
Solubility Information Insoluble in water.
Formula Weight 59.91
Density 4.93 g/mL
Chemical Name or Material Titanium carbide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'