Learn More
Titanium carbide, nanopowder, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 12070-08-5 |
---|---|
Molecular Formula | CTi |
Molecular Weight (g/mol) | 59.88 |
MDL Number | MFCD00011268 |
InChI Key | YXIVWSJCLXKLJL-UHFFFAOYSA-N |
IUPAC Name | methanidylidynetitaniumylium |
SMILES | [C-]#[Ti+] |
Description
Titanium carbide is used in the manufacture of wear-resistant tools, cutting tools, As a coating for abrasive steel bearings, In the form of nano titanium carbide ceramic in optics applications, Enhance the conductivity of materials and as a nucleating agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12070-08-5 | |
59.88 | |
YXIVWSJCLXKLJL-UHFFFAOYSA-N | |
[C-]#[Ti+] |
CTi | |
MFCD00011268 | |
methanidylidynetitaniumylium |
Specifications
4820°C | |
12070-08-5 | |
CTi | |
Soluble in nitric acid and aqua regia. Insoluble in water. | |
[C-]#[Ti+] | |
59.88 | |
4.93 |
∽3140°C | |
Powder | |
MFCD00011268 | |
YXIVWSJCLXKLJL-UHFFFAOYSA-N | |
methanidylidynetitaniumylium | |
59.91 | |
Titanium carbide |
Safety and Handling
P210-P240-P241-P280-P370+P378q
H228
missing translation for 'dotInformation' : Hazard Class: 4.1; Packaging Group: III
missing translation for 'einecsNumber' : 235-120-4
missing translation for 'rtecsNumber' : XR1903500
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only