missing translation for 'onlineSavingsMsg'
Learn More

Titanium foil, 0.5mm (0.02in) thick, annealed, 99% (metals basis), Thermo Scientific Chemicals

Product Code. 11385907
Click to view available options
Quantity:
100 x 100 mm
200 x 200 mm
200 x 1000 mm
This item is not returnable. View return policy
This item is not returnable. View return policy

Form: approx. 22.5g/100x100mm

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-32-6
Molecular Formula Ti
Molecular Weight (g/mol) 47.87
MDL Number MFCD00011264,MFCD00151301
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
Synonym oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
PubChem CID 23963
ChEBI CHEBI:33341
IUPAC Name titanium
SMILES [Ti]

Specifications

Color Silver to Gray
Appearance Silver Gray foil
Assay Percent Range 99% (Metals basis)
Quantity 100 x 100 mm
Odor Odorless
Form Foil
Chemical Name or Material Titanium, Annealed

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.