missing translation for 'onlineSavingsMsg'
Learn More

Titanium Granules, -15 mesh, 99.9% (Metals basis excluding Na and K), Thermo Scientific™

Product Code. 11341519
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-32-6
Molecular Formula Ti
Molecular Weight (g/mol) 47.87
MDL Number MFCD00011264,MFCD00151301
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
Synonym oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
PubChem CID 23963
ChEBI CHEBI:33341
IUPAC Name titanium
SMILES [Ti]

Specifications

Color Gray
Appearance Dark Gray granules
Density 4.506g/cm3
Assay Percent Range 99.9% (Metals basis excluding Na and K)
Quantity 250 g
Mesh Size -15 Mesh
Odor Odorless
Form Granular
Chemical Name or Material Titanium
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'