missing translation for 'onlineSavingsMsg'
Learn More

Titanium(II) oxide pellets, 99.9% (metals basis), Thermo Scientific Chemicals

168.84 € - 499.49 €

Chemical Identifiers

CAS 12137-20-1
Molecular Formula OTi
Molecular Weight (g/mol) 63.87
MDL Number MFCD00036279
InChI Key OGIDPMRJRNCKJF-UHFFFAOYSA-N
Synonym titanium monoxide, titanium ii oxide, titanium oxide tio, oxido titanium, titanium ii oxide pellets, titanium oxide, 99.9 %, titanium ii oxide, optical grade, titanium ii oxide trace metals basis 5g, titanium ii oxide, coating quality, granular, titanium ii oxide, pieces trace metals basis
PubChem CID 61685
IUPAC Name oxotitanium
SMILES O=[Ti]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11350208
View Documents
Thermo Scientific Alfa Aesar
042940.14
25 g
168.84 €
25g
Please to purchase this item. Need a web account? Register with us today!
11360208
View Documents
Thermo Scientific Alfa Aesar
042940.22
100 g
499.49 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Used for vacuum deposition. Also applications include sunscreens and UV-blocking in cosmetics, varnishes, wood preservatives, textile fibers, and packaging films. Also used as paint pigment, opacifying agent, welding rod fluxes, optical coatings, as a catalyst, ceramic finish coat, plastics, elastomers, coated fabrics, printing inks, roofing granules, glass, and in glazes. Also identified as Titanium monoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12137-20-1
63.87
OGIDPMRJRNCKJF-UHFFFAOYSA-N
61685
O=[Ti]
OTi
MFCD00036279
titanium monoxide, titanium ii oxide, titanium oxide tio, oxido titanium, titanium ii oxide pellets, titanium oxide, 99.9 %, titanium ii oxide, optical grade, titanium ii oxide trace metals basis 5g, titanium ii oxide, coating quality, granular, titanium ii oxide, pieces trace metals basis
oxotitanium

Specifications

>3000°C
12137-20-1
MFCD00036279
Insoluble in water.
O=[Ti]
63.87
63.9
Odorless
(metals basis)
1700°C
OTi
titanium monoxide, titanium ii oxide, titanium oxide tio, oxido titanium, titanium ii oxide pellets, titanium oxide, 99.9 %, titanium ii oxide, optical grade, titanium ii oxide trace metals basis 5g, titanium ii oxide, coating quality, granular, titanium ii oxide, pieces trace metals basis
OGIDPMRJRNCKJF-UHFFFAOYSA-N
oxotitanium
61685
99.9%
4.95 g/mL
Titanium (II) oxide pellets, 8 to 9mm(0.32 to 0.36 in.) dia. x 6 to 7mm(0.24 to 0.28 in.) thick
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 235-236-5

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only