missing translation for 'onlineSavingsMsg'
Learn More

Titanium(III) chloride, 20% w/v solution in 2N Hydrochloric acid, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 395622500

97.60 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.



 View more versions of this product

Product Code. 10453935

  • 98.09 € / 250mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Titanium(III) chloride
Purple
250 mL
TiCl3
15,9637
Solubility in water: soluble. Other solubilities: soluble in alcohol, practically insoluble in ether
[Cl-].[Cl-].[Cl-].[Ti+3]
154.22
∼20% (TiCl3) (exact strength stated on certificate of analysis)
1.2000g/mL
7705-07-9, 7647-01-0, 7732-18-5
Liquid
Cl3Ti
MFCD00011266
Titanous chloride
YONPGGFAJWQGJC-UHFFFAOYSA-K
titanium(3+) trichloride
154.26
Plastic bottle
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Titanium(III) chloride, 20% w/v solution in 2N Hydrochloric acid

Signal Word
  • Danger

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.

Supplemental information
  • MIXTURE LIST-Contains : Titanium trichloride, Hydrochloric acid,Phosphoric acid

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only