missing translation for 'onlineSavingsMsg'
Learn More

Titanium(III) oxide, 99.8% (metals basis), Thermo Scientific Chemicals

178.89 € - 695.46 €

Chemical Identifiers

CAS 1344-54-3
Molecular Formula O3Ti2
Molecular Weight (g/mol) 143.731
MDL Number MFCD00036278
InChI Key GQUJEMVIKWQAEH-UHFFFAOYSA-N
Synonym titanium iii oxide, oxo oxotitaniooxy titanium, oxo-oxotitaniooxy titanium, titanium sesquioxide, titatnum iii oxide, ti2o3 nanoparticle, ti2o3, titanium oxide ti2o3, titanium iii oxide nanoparticle, titanium iii oxide trace metals basis 50g
PubChem CID 123111
IUPAC Name oxo(oxotitaniooxy)titanium
SMILES O=[Ti]O[Ti]=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11364889
View Documents Promotion Details
Thermo Scientific Alfa Aesar
077127.18
50 g
178.89 €
50g
Please to purchase this item. Need a web account? Register with us today!
11374889
View Documents Promotion Details
Thermo Scientific Alfa Aesar
077127.30
250 g
695.46 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Titanium(III) oxide is used to study the bioacceptability of medical implant surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1344-54-3
143.731
GQUJEMVIKWQAEH-UHFFFAOYSA-N
123111
O=[Ti]O[Ti]=O
O3Ti2
MFCD00036278
titanium iii oxide, oxo oxotitaniooxy titanium, oxo-oxotitaniooxy titanium, titanium sesquioxide, titatnum iii oxide, ti2o3 nanoparticle, ti2o3, titanium oxide ti2o3, titanium iii oxide nanoparticle, titanium iii oxide trace metals basis 50g
oxo(oxotitaniooxy)titanium

Specifications

2130°C (decomposition)
Powder
MFCD00036278
Insoluble in water.
O=[Ti]O[Ti]=O
143.731
143.8
Odorless
(metals basis)
1344-54-3
O3Ti2
titanium iii oxide, oxo oxotitaniooxy titanium, oxo-oxotitaniooxy titanium, titanium sesquioxide, titatnum iii oxide, ti2o3 nanoparticle, ti2o3, titanium oxide ti2o3, titanium iii oxide nanoparticle, titanium iii oxide trace metals basis 50g
GQUJEMVIKWQAEH-UHFFFAOYSA-N
oxo(oxotitaniooxy)titanium
123111
99.8%
4.6 g/mL
Titanium (III) oxide
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 215-697-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only