Learn More
Titanium(III) sulfate, 20% in 1-4% sulfuric acid
Description
Titanium(III) sulfate is used in the manufacture of bulk, large scale chemicals (including petroleum products), fine chemicals, basic metals, including alloys, computer, electronic and optical products, electrical equipment. It is also used as a biochemical for proteomics research. It is a chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Liquid |
| Quantity | 1 kg |
| UN Number | UN3264 |
| Merck Index | 14,9476 |
| Solubility Information | Fully miscible with water. |
| Formula Weight | 383.38 |
| Concentration or Composition (by Analyte or Components) | 20% in 1 to 4% sulfuric acid |
| Density | 1.15 g/mL |
| Chemical Name or Material | Titanium(III) sulfate |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.