missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) bromide, 98%, Thermo Scientific Chemicals

74.37 € - 258.06 €

Chemical Identifiers

CAS 7789-68-6
Molecular Formula TiBr4
MDL Number MFCD00011265
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11364507
View Documents
Thermo Scientific Alfa Aesar
039592.09
10 g
74.37 €
10g
Please to purchase this item. Need a web account? Register with us today!
11374507
View Documents
Thermo Scientific Alfa Aesar
039592.18
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Titanium(IV) bromide is used as a Lewis acid catalyst in organic synthesis. It reacts with tetrahydrofurnan and 2-methylpyridine and forms adducts such as TiBr4(THF)2 and TiBr4(2-MePy). It is also used for the formation of titanium silicide thin films on silicon substrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7789-68-6
TiBr4
MFCD00011265

Specifications

233°C
7789-68-6
Crystalline
TiBr4
Soluble in absolute alcohol. Decomposes in waterSoluble in chlorocarbons,benzene and alcohol.
Acrid
2.6
39°C
Orange
98%
MFCD00011265
367.54
Moisture sensitive
Titanium(IV) bromide
Videos
Safety and Handling

Safety and Handling

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II

missing translation for 'einecsNumber' : 232-185-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.