missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) bromide, 98%, Thermo Scientific Chemicals

Product Code. 10591314
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-68-6
Molecular Formula TiBr4
Molecular Weight (g/mol) 367.5
MDL Number MFCD00011265

Specifications

Boiling Point 230.0°C
Melting Point 39.0°C
Quantity 100 g
Assay Percent Range 97.5% min. (Argentometry)
Linear Formula TiBr4
Merck Index 15,9633
Solubility Information Solubility in water: reacts. Other solubilities: soluble in absolute ether and absolute alcohol
Formula Weight 367.5
Percent Purity 98%
Packaging Glass bottle
Chemical Name or Material Titanium(IV) bromide
Show More Show Less
compliance-icons
Product Identifier
  • Titanium(IV) bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'