missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) bromide, 98%, Thermo Scientific Chemicals

42.71 € - 402.00 €

Chemical Identifiers

CAS 7789-68-6
Molecular Formula TiBr4
Molecular Weight (g/mol) 367.5
MDL Number MFCD00011265
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10771525
SDS View Documents
Thermo Scientific Acros
391240050
5 g
42.71 €
5g
Please to purchase this item. Need a web account? Register with us today!
10085143
SDS View Documents
Thermo Scientific Acros
391240250
25 g
123.62 €
25g
Please to purchase this item. Need a web account? Register with us today!
10591314
SDS View Documents
Thermo Scientific Acros
391241000
100 g
402.00 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7789-68-6
TiBr4
367.5
MFCD00011265

Specifications

230.0°C
7789-68-6
TiBr4
MFCD00011265
Solubility in water: reacts. Other solubilities: soluble in absolute ether and absolute alcohol
367.5
Glass bottle
39.0°C
97.5% min. (Argentometry)
TiBr4
15,9633
367.5
98%
Titanium(IV) bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 232-185-0

Product Identifier
  • Titanium(IV) bromide

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.