missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) chloride, 1M solution in dichloromethane, AcroSeal™, Thermo Scientific Chemicals

93.80 € - 262.00 €

Specifications

Chemical Name or Material Titanium(IV) chloride
Name Note 1M Solution in Dichloromethane
CAS 7550-45-0, 75-09-2
Color Brown to Yellow
Physical Form Solution
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10575443
View Documents Promotion Details
Thermo Scientific Acros
377741000
100 mL AcroSeal™ Glass bottle
93.80 €
100mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10086522
View Documents Promotion Details
Thermo Scientific Acros
377748000
800 mL AcroSeal™ Glass Bottle
262.00 €
800mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
Specifications

Specifications

Titanium(IV) chloride
7550-45-0, 75-09-2
Solution
MFCD00011267
titanium iv chloride, titanium 4+ tetrachloride, acmc-1bjgv, titanium 4+ ion tetrachloride, parent, titanium +4 cation tetrachloride, titanium iv chloride 100ml, superlist names titanium chloride, t-4, titanic chloride; titanio tetracloruro di, unii-8o3pje5t7q; titanium chloride ticl4
XJDNKRIXUMDJCW-UHFFFAOYSA-J
titanium(4+);tetrachloride
160960
0.95 to 1.10M
1.3650g/mL
1M Solution in Dichloromethane
Brown to Yellow
Cl4Ti
15, 9634
Solubility in water: reacts
[Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]
189.71
189.71
1.365
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Suspected of causing cancer if inhaled.
Toxic if inhaled.
May cause drowsiness or dizziness.
May cause respiratory irritation.
Reacts violently with water.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 231-441-9

Product Identifier
  • Titanium(IV) chloride, 1M solution in dichloromethane

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity after single exposure Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H370-Causes damage to organs.
  • H372-Causes damage to organs.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H336-May cause drowsiness or dizziness.
  • H331-Toxic if inhaled.
  • EUH014-Reacts violently with water.
  • EUH071-Corrosive to the respiratory tract.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only