Learn More
Titanium(IV) chloride, 1M solution in toluene, AcroSeal™, Thermo Scientific Chemicals
Specifications
Chemical Name or Material | Titanium(IV) chloride |
---|---|
Name Note | 1M Solution in Toluene |
CAS | 7550-45-0, 108-88-3 |
Color | Orange |
Physical Form | Liquid |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidSpecifications
Titanium(IV) chloride | |
7550-45-0, 108-88-3 | |
Liquid | |
MFCD00011267 | |
15, 9634 | |
Solubility in water: reacts | |
[Cl-].[Cl-].[Cl-].[Cl-].[Ti+4] | |
189.71 | |
189.71 | |
0.964 |
1M Solution in Toluene | |
Orange | |
Cl4Ti | |
8°C | |
titanium iv chloride, titanium 4+ tetrachloride, acmc-1bjgv, titanium 4+ ion tetrachloride, parent, titanium +4 cation tetrachloride, titanium iv chloride 100ml, superlist names titanium chloride, t-4, titanic chloride; titanio tetracloruro di, unii-8o3pje5t7q; titanium chloride ticl4 | |
XJDNKRIXUMDJCW-UHFFFAOYSA-J | |
titanium(4+);tetrachloride | |
160960 | |
0.95 to 1.1M | |
0.9640g/mL |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Toxic if inhaled.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Suspected of damaging the unborn child.
May cause drowsiness or dizziness
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for seve
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-441-9
- Titanium(IV) chloride, 1M solution in toluene
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 1
- Specific target organ toxicity after single exposure Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H318-Causes serious eye damage.
- H370-Causes damage to organs.
- H372-Causes damage to organs.
- H314-Causes severe skin burns and eye damage.
- H361d-Suspected of damaging the unborn child.
- H331-Toxic if inhaled.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
- EUH071-Corrosive to the respiratory tract.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only