missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) iodide, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11378588
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Titanium(IV) iodide, anhydrous is used in the CVD of titanium nitride films with optimized characteristics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7720-83-4
MDL Number MFCD00043077

Specifications

Melting Point 150°C
Physical Form Lump
Quantity 10 g
UN Number UN3260
Solubility Information Soluble in dichloromethane,chloroform,carbon disulfide.
Formula Weight 555.52
Percent Purity 99.9%
Odor Acrid
Sensitivity Moisture sensitive
Density 4.30 g/mL
Assay (metals basis)
Chemical Name or Material Titanium(IV) iodide, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.