missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) isobutoxide, Thermo Scientific Chemicals

40.80 € - 402.00 €

Chemical Identifiers

CAS 7425-80-1
MDL Number MFCD00015045
Synonym tetrakis 2-methylpropyl orthosilicate, titanium iv isobutoxide, tetrakis 2-methylpropyl silicate, tetraisobutyl orthosilicate, silane, tetraisobutoxy, silicic acid tetraisobutyl ester, orthosilicic acid tetrakis 2-methylpropyl ester, silicic acid h4sio4 ,tetrakis 2-methylpropyl ester
PubChem CID 69626
IUPAC Name tetrakis(2-methylpropyl) silicate
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15413007
SDS View Documents
Thermo Scientific Alfa Aesar
089455.18
50 g
40.80 €
50g
Please to purchase this item. Need a web account? Register with us today!
15423007
SDS View Documents
Thermo Scientific Alfa Aesar
089455.30
250 g
111.56 €
250g
Please to purchase this item. Need a web account? Register with us today!
15433007
SDS View Documents
Thermo Scientific Alfa Aesar
089455.A1
1 kg
402.00 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Titanium(IV) isobutoxide general application in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7425-80-1
tetrakis 2-methylpropyl orthosilicate, titanium iv isobutoxide, tetrakis 2-methylpropyl silicate, tetraisobutyl orthosilicate, silane, tetraisobutoxy, silicic acid tetraisobutyl ester, orthosilicic acid tetrakis 2-methylpropyl ester, silicic acid h4sio4 ,tetrakis 2-methylpropyl ester
tetrakis(2-methylpropyl) silicate
MFCD00015045
69626

Specifications

296°C
Yellow
C16H36O4Si
MFCD00015045
tetrakis 2-methylpropyl orthosilicate, titanium iv isobutoxide, tetrakis 2-methylpropyl silicate, tetraisobutyl orthosilicate, silane, tetraisobutoxy, silicic acid tetraisobutyl ester, orthosilicic acid tetrakis 2-methylpropyl ester, silicic acid h4sio4 ,tetrakis 2-methylpropyl ester
QKCMIPQMYBKEPP-UHFFFAOYSA-N
tetrakis(2-methylpropyl) silicate
69626
1.495
1.02 g/mL
7425-80-1
Melt
Ti[(CH3)2CHCH2O]4
77°C (171°F)
Reacts with water.
CC(C)CO[Si](OCC(C)C)(OCC(C)C)OCC(C)C
320.545
340.34
Moisture sensitive
Titanium (IV) isobutoxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

P264b-P280i-P305+P351+P338

H319

missing translation for 'einecsNumber' : 231-061-3

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

Product Identifier
  • Titanium(IV) isobutoxide

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Titanium(IV) isobutoxide

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.