missing translation for 'onlineSavingsMsg'
Learn More
Titanium(IV) isopropoxide, 97+%, Thermo Scientific Chemicals Product Code.: 15442957

Titanium(IV) isopropoxide, 97+%, Thermo Scientific Chemicals

Product Code. 15442957
500 g
Click to view available options
Quantity:
10 g
100 g
500 g
4 x 500 g
This item is not returnable. View return policy

Product Code. 15442957

Brand: Thermo Scientific Alfa Aesar 077115.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Catalyzes the regio selective ring opening of 2,3 epoxy alcohols and acids

Catalyzes the regio selective ring opening of 2,3 epoxy alcohols and acids.Titanium(IV) isopropoxide is used as a precursor for the preparation of titanium and barium-strontium-titanate thin films. It is useful to make porous titanosilicates and potential ion-exchange materials for cleanup of radioactive wastes. It is an active component of sharpless epoxidation as well as involved in the synthesis of chiral epoxides. In Kulinkovich reaction, it is involved as a catalyst in the preparation of cyclopropanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 546-68-9
Molecular Formula C12H28O4Ti
Molecular Weight (g/mol) 284.219
MDL Number MFCD00008871
InChI Key VXUYXOFXAQZZMF-UHFFFAOYSA-N
Synonym titanium tetraisopropanolate, titanium iv isopropoxide, tetraisopropyl orthotitanate, titanium isopropoxide, titanium tetraisopropylate, titanium isopropylate, ti isopropylate, tetraisopropoxytitanium iv, isopropyl orthotitanate, tetraisopropyl titanate
PubChem CID 11026
IUPAC Name propan-2-olate;titanium(4+)
SMILES CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-].[Ti+4]

Specifications

Boiling Point 232°C
Melting Point 16°C to 20°C
Color Colorless to Yellow
Physical Form Liquid
Quantity 500 g
Linear Formula Ti[OCH(CH3)2]4
UN Number UN2413
Beilstein 3679474
Flash Point 46°C (115°F)
Merck Index 14,9480
Solubility Information Soluble in anhydrous ethanol, ether, benzene and chloroform; Decomposes in water
Formula Weight 284.23
Percent Purity ≥97%
Refractive Index 1.464
Sensitivity Moisture sensitive
Density 0.955 g/mL
Chemical Name or Material Titanium (IV) isopropoxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number62500192 Certificate TypeCertificate of Analysis Date10/04/2025 Catalog Number077115.09,077115.36,077115.22,077115.57
Lot Number10253534 Certificate TypeCertificate of Analysis Date19/12/2024 Catalog Number077115.09,077115.36,077115.22,077115.57
Lot NumberX29K014 Certificate TypeCertificate of Analysis Date07/11/2024 Catalog Number077115.09,077115.36,077115.22,077115.57
Lot NumberS12K028 Certificate TypeCertificate of Analysis Date19/06/2024 Catalog Number077115.09,077115.36,077115.22,077115.57
Lot NumberY21J007 Certificate TypeCertificate of Analysis Date04/01/2024 Catalog Number077115.09,077115.36,077115.22,077115.57

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Titanium(IV) isopropoxide, 97+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.