missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) methoxide, 95%, Thermo Scientific Chemicals

Product Code. 11334899
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Titanium(IV) methoxide is used for the preparation of polyoxotitanates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 992-92-7
Molecular Formula Ti(OCH3)4·(CH3OH)x, x<1
MDL Number MFCD00014883
Synonym tetramethyl titanate, titanium tetramethoxide, titanium iv methoxide, methanolate; titanium 4+, titanium tetra methanolate, methanol, titanium 4+ salt, titanium methoxide, methanol, titanium 4+ salt 4:1, titanium 4+ tetrakis methoxide, titanium 4+ ion tetrakis methoxide

Specifications

Boiling Point >243°C (52mmHg)
Melting Point 200°C to 210°C
Physical Form Granular/Lump
Quantity 25 g
UN Number UN3181
Flash Point 48°C (118°F)
Solubility Information Slightly soluble in organic solvents.
Percent Purity 95%
Sensitivity Moisture sensitive
Chemical Name or Material Titanium (IV) methoxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'