missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium(IV) oxide, 20-36% in H{2}O colloidal dispersion, Thermo Scientific Chemicals
UV protection in clear coatings
Chemical Identifiers
CAS | 13463-67-7 |
---|---|
Molecular Formula | O2Ti |
Molecular Weight (g/mol) | 79.87 |
MDL Number | MFCD00011269 MFCD00210650 |
InChI Key | GWEVSGVZZGPLCZ-UHFFFAOYSA-N |
Synonym | titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance |
PubChem CID | 26042 |
ChEBI | CHEBI:32234 |
IUPAC Name | dioxotitanium |
SMILES | O=[Ti]=O |
Description
UV protection in clear coatings. Photo electrodes of varying properties can be made using colloidal suspensions of titanium(IV) oxide nanoparticles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13463-67-7 | |
79.87 | |
GWEVSGVZZGPLCZ-UHFFFAOYSA-N | |
26042 | |
dioxotitanium |
O2Ti | |
MFCD00011269 MFCD00210650 | |
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance | |
CHEBI:32234 | |
O=[Ti]=O |
Specifications
368°C | |
O2Ti | |
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance | |
GWEVSGVZZGPLCZ-UHFFFAOYSA-N | |
dioxotitanium | |
26042 | |
79.9 | |
1.26 g/mL |
13463-67-7 | |
MFCD00011269 MFCD00210650 | |
Insoluble in water. | |
O=[Ti]=O | |
79.87 | |
CHEBI:32234 | |
20 to 35% in H2O | |
Titanium (IV) oxide, Colloidal dispersion |
Safety and Handling
GHS H Statement
H351
Suspected of causing cancer.
P201-P202-P281-P308+P313-P501c
H351
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only