missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) oxide bis(2,4-pentanedionate), Thermo Scientific Chemicals

100.00 € - 2027.00 €

Chemical Identifiers

CAS 14024-64-7
Molecular Formula C10H16O5Ti
Molecular Weight (g/mol) 264.1
MDL Number MFCD00013505
InChI Key ADVORQMAWLEPOI-UHFFFAOYSA-N
Synonym bis acetylacetonato titanium iv oxide, bis 4-hydroxypent-3-en-2-one ; titanium ii oxide
PubChem CID 74765597
IUPAC Name 4-hydroxypent-3-en-2-one;oxotitanium
SMILES CC(=CC(=O)C)O.CC(=CC(=O)C)O.O=[Ti]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11384889
View Documents Promotion Details
Thermo Scientific Alfa Aesar
077130.09
10 g
100.00 €
10g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11304899
View Documents Promotion Details
Thermo Scientific Alfa Aesar
077130.18
50 g
464.00 €
50g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11394889
View Documents Promotion Details
Thermo Scientific Alfa Aesar
077130.30
250 g
2027.00 €
250g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Employed as a cross linking agent for cellulose fibers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14024-64-7
264.1
ADVORQMAWLEPOI-UHFFFAOYSA-N
74765597
CC(=CC(=O)C)O.CC(=CC(=O)C)O.O=[Ti]
C10H16O5Ti
MFCD00013505
bis acetylacetonato titanium iv oxide, bis 4-hydroxypent-3-en-2-one ; titanium ii oxide
4-hydroxypent-3-en-2-one;oxotitanium

Specifications

184°C (decomposition)
Powder
MFCD00013505
Soluble in water (6.6g/L (20°C)).
CC(=CC(=O)C)O.CC(=CC(=O)C)O.O=[Ti]
264.1
262.12
14024-64-7
C10H16O5Ti
bis acetylacetonato titanium iv oxide, bis 4-hydroxypent-3-en-2-one ; titanium ii oxide
ADVORQMAWLEPOI-UHFFFAOYSA-N
4-hydroxypent-3-en-2-one;oxotitanium
74765597
Titanium (IV) oxide bis (2,4-pentanedionate)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'einecsNumber' : 237-861-9

missing translation for 'rtecsNumber' : XR2330000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only