Learn More
Titanium(IV) oxide, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
105.53 € - 1086.41 €
Chemical Identifiers
CAS | 13463-67-7 |
---|---|
Molecular Formula | O2Ti |
Molecular Weight (g/mol) | 79.87 |
MDL Number | MFCD00011269 MFCD00210650 |
InChI Key | GWEVSGVZZGPLCZ-UHFFFAOYSA-N |
Synonym | titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance |
PubChem CID | 26042 |
ChEBI | CHEBI:32234 |
IUPAC Name | dioxotitanium |
SMILES | O=[Ti]=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11307077
|
Thermo Scientific Alfa Aesar
010897.09 |
10 g |
105.53 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11397067
|
Thermo Scientific Alfa Aesar
010897.18 |
50 g |
355.77 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11387067
|
Thermo Scientific Alfa Aesar
010897.30 |
250 g |
1086.41 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Titanium(IV) oxide is used in paints, varnishes, paper, plastics printing inks, sun screen, fibers, rubber, cosmetic products, anti-fogging coatings, foodstuffs, pure titanium, glass, glass ceramics, memristor, electrical ceramics, catalysts, electric conductors and chemical intermediates. It exhibit excellent photocatalytic properties (Honda-Fujishima effect) In addition, titanium dioxide is also used to cleave protein at the site where proline is present.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13463-67-7 | |
79.87 | |
GWEVSGVZZGPLCZ-UHFFFAOYSA-N | |
26042 | |
dioxotitanium |
O2Ti | |
MFCD00011269 MFCD00210650 | |
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance | |
CHEBI:32234 | |
O=[Ti]=O |
Specifications
2500°C to 3000°C | |
13463-67-7 | |
99.995% (metals basis) | |
MFCD00011269 MFCD00210650 | |
Slightly soluble in hydrofluoric acid and hot concentrated sulfuric acid. Insoluble in water,hydrochloric acid,dilute sulfuric acid,and organic solvents. | |
O=[Ti]=O | |
79.87 | |
CHEBI:32234 | |
4.26 |
1830°C to 1850°C | |
Powder | |
O2Ti | |
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance | |
GWEVSGVZZGPLCZ-UHFFFAOYSA-N | |
dioxotitanium | |
26042 | |
79.9 | |
Titanium(IV) oxide |
Safety and Handling
P201-P202-P281-P308+P313-P501c
H351
missing translation for 'einecsNumber' : 236-675-5
missing translation for 'rtecsNumber' : XR2275000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only