missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) oxide, Puriss., meets analytical spec. of BP, Ph. Eur., USP, 99 to 100.5%, Honeywell Fluka™
BIOPHARMA

Product Code. 15641390
Click to view available options
Quantity:
250 g
1 kg
25 kg
6 x 1 kg

Puriss., meets analytical specification of BP, Ph. Eur., USP, 99-100.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13463-67-7
Molecular Formula O2Ti
Molecular Weight (g/mol) 79.87
MDL Number MFCD00011269 MFCD00210650
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
Synonym titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
PubChem CID 26042
ChEBI CHEBI:32234
IUPAC Name dioxotitanium
SMILES O=[Ti]=O

Specifications

Quantity 250 g
Linear Formula TiO2
UN Number NONH for all modes of transport
Formula Weight 79.87g/mol
Percent Purity 99 to 100.5%
Grade puriss.
Density 4.26g/mL at 25°C
Chemical Name or Material Titanium(Iv) Oxide, Meets analytical specification of Ph. Eur.BP, USP
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'