Learn More
Titanium(IV) oxide sulfate sulfuric acid hydrate, Thermo Scientific Chemicals
98.99 € - 304.52 €
Chemical Identifiers
CAS | 123334-00-9 |
---|---|
Molecular Formula | TiOSO4.xH2O + H2SO4 |
MDL Number | MFCD00011270 |
Synonym | Titanyl sulfate, Titanium(IV) oxysulfate-sulfuric acid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11354507
|
Thermo Scientific Alfa Aesar
039591.36 |
500 g |
98.99 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11344507
|
Thermo Scientific Alfa Aesar
039591.A3 |
2 kg |
304.52 €
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Titanium(IV) oxide sulfate sulfuric acid hydrate is widely used as a pigment in paints and dyes. It is also useful in the preparation of stable amorphous titanium oxide hollow microspheres.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
123334-00-9 | |
TiOSO4.xH2O + H2SO4 | |
MFCD00011270 | |
Titanyl sulfate, Titanium(IV) oxysulfate-sulfuric acid |
Specifications
123334-00-9 | |
Powder | |
MFCD00011270 | |
Soluble in water. |
White | |
TiOSO4.xH2O + H2SO4 | |
Titanyl sulfate, Titanium(IV) oxysulfate-sulfuric acid | |
Titanium(IV) oxide sulfate sulfuric acid hydrate |
Safety and Handling
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only