missing translation for 'onlineSavingsMsg'
Learn More

Titanium nitride, 99.7% (metals basis), Thermo Scientific Chemicals

96.58 € - 938.67 €

Chemical Identifiers

CAS 25583-20-4
Molecular Formula NTi
Molecular Weight (g/mol) 61.87
MDL Number MFCD00049596
InChI Key NRTOMJZYCJJWKI-UHFFFAOYSA-N
Synonym titanium nitride, titanium nitride tin, nitridotitanium, unii-6rw464feff, titannitrid, titaniumylidyneamine, titanium mononitride, titanium nitride, <3 mum, 6rw464feff, titanium nitride tin grade a
PubChem CID 93091
IUPAC Name azanylidynetitanium
SMILES N#[Ti]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11382458
View Documents Promotion Details
Thermo Scientific Alfa Aesar
014510.18
50 g
96.58 €
50g
Please to purchase this item. Need a web account? Register with us today!
11372458
View Documents Promotion Details
Thermo Scientific Alfa Aesar
014510.30
250 g
318.59 €
250g
Please to purchase this item. Need a web account? Register with us today!
11392458
View Documents Promotion Details
Thermo Scientific Alfa Aesar
014510.A1
1 kg
938.67 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Titanium nitride is a hard ceramic material used as a coating on titanium alloys, steel, carbide and aluminum components. It finds application in jewelry and high-performance ceramic materials due to its gold colored surface and in medical devices such as scalpel blades and orthopedic bone saw blades. Further, it is used as a conductive barrier in microelectronics and electrodes in bioelectronic applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

25583-20-4
61.87
NRTOMJZYCJJWKI-UHFFFAOYSA-N
93091
N#[Ti]
NTi
MFCD00049596
titanium nitride, titanium nitride tin, nitridotitanium, unii-6rw464feff, titannitrid, titaniumylidyneamine, titanium mononitride, titanium nitride, <3 mum, 6rw464feff, titanium nitride tin grade a
azanylidynetitanium

Specifications

2950°C
Powder
NTi
titanium nitride, titanium nitride tin, nitridotitanium, unii-6rw464feff, titannitrid, titaniumylidyneamine, titanium mononitride, titanium nitride, <3 mum, 6rw464feff, titanium nitride tin grade a
NRTOMJZYCJJWKI-UHFFFAOYSA-N
azanylidynetitanium
93091
5.22
25583-20-4
99.7% (metal basis)
MFCD00049596
Insoluble in water.
N#[Ti]
61.87
61.91
Titanium nitride
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 247-117-5

missing translation for 'rtecsNumber' : XR2230000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only