missing translation for 'onlineSavingsMsg'
Learn More
Titanium rod, 12.7mm (0.5in) dia, annealed, 99.5% (metals basis), Thermo Scientific Chemicals Product Code.: 11325868

Titanium rod, 12.7mm (0.5in) dia, annealed, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11325868
10 cm
Click to view available options
Quantity:
10 cm
50 cm
250 cm
This item is not returnable. View return policy

Product Code. 11325868

Brand: Thermo Scientific Alfa Aesar 010394.BS

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: approx. 5.7g/cm

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-32-6
Molecular Formula Ti
Molecular Weight (g/mol) 47.87
MDL Number MFCD00011264,MFCD00151301
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
Synonym oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
PubChem CID 23963
ChEBI CHEBI:33341
IUPAC Name titanium
SMILES [Ti]

Specifications

Color Silver to Gray
Appearance Silver Gray rod
Assay Percent Range 99.5% (Metals basis)
Quantity 10 cm
Odor Odorless
Form Rod
Chemical Name or Material Titanium, Annealed

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY22K003 Certificate TypeCertificate of Analysis Date25/11/2024 Catalog Number010394.CB,010394.BW,010394.BS
Lot NumberY22K002 Certificate TypeCertificate of Analysis Date25/11/2024 Catalog Number010394.CB,010394.BW,010394.BS
Lot NumberP28I055 Certificate TypeCertificate of Analysis Date19/08/2022 Catalog Number010394.CB,010394.BW,010394.BS
Lot NumberZ18A023 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number010394.CB,010394.BW,010394.BS
Lot NumberW10H010 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number010394.CB,010394.BW,010394.BS

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Titanium rod, 12.7mm (0.5in) dia, annealed, 99.5% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.