Learn More
Titanium silicide, 99.5% (metals basis), Thermo Scientific Chemicals
167.00 € - 544.00 €
Chemical Identifiers
CAS | 12039-83-7 |
---|---|
Molecular Formula | Si2Ti |
Molecular Weight (g/mol) | 104.037 |
MDL Number | MFCD00151541 |
InChI Key | DFJQEGUNXWZVAH-UHFFFAOYSA-N |
Synonym | titanium silicide, titanium disilicide, titanium silicide tisi2, titanium silicide, tisi, titanium silicide sputtering target, 50.8mm 2.0in dia x 3.18mm 0.125in thick, titanium silicide sputtering target, 50.8mm 2.0in dia x 6.35mm 0.250in thick, titanium silicide sputtering target, 76.2mm 3.0in dia x 3.18mm 0.125in thick, titanium silicide sputtering target, 76.2mm 3.0in dia x 6.35mm 0.250in thick |
PubChem CID | 6336889 |
IUPAC Name | bis($l^{2}-silanylidene)titanium |
SMILES | [Si]=[Ti]=[Si] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11382178
|
Thermo Scientific Alfa Aesar
013088.18 |
50 g |
167.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11392178
|
Thermo Scientific Alfa Aesar
013088.30 |
250 g |
544.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Titanium silicide is used in the semiconductor industry. It is also used in the fabrication of transistors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12039-83-7 | |
104.037 | |
DFJQEGUNXWZVAH-UHFFFAOYSA-N | |
6336889 | |
[Si]=[Ti]=[Si] |
Si2Ti | |
MFCD00151541 | |
titanium silicide, titanium disilicide, titanium silicide tisi2, titanium silicide, tisi, titanium silicide sputtering target, 50.8mm 2.0in dia x 3.18mm 0.125in thick, titanium silicide sputtering target, 50.8mm 2.0in dia x 6.35mm 0.250in thick, titanium silicide sputtering target, 76.2mm 3.0in dia x 3.18mm 0.125in thick, titanium silicide sputtering target, 76.2mm 3.0in dia x 6.35mm 0.250in thick | |
bis($l^{2}-silanylidene)titanium |
Specifications
1760°C | |
Gray | |
Si2Ti | |
MFCD00151541 | |
titanium silicide, titanium disilicide, titanium silicide tisi2, titanium silicide, tisi, titanium silicide sputtering target, 50.8mm 2.0in dia x 3.18mm 0.125in thick, titanium silicide sputtering target, 50.8mm 2.0in dia x 6.35mm 0.250in thick, titanium silicide sputtering target, 76.2mm 3.0in dia x 3.18mm 0.125in thick, titanium silicide sputtering target, 76.2mm 3.0in dia x 6.35mm 0.250in thick | |
DFJQEGUNXWZVAH-UHFFFAOYSA-N | |
bis($l^{2}-silanylidene)titanium | |
6336889 | |
99.5% | |
Titanium silicide |
12039-83-7 | |
Powder | |
TiSi2 | |
UN3178 | |
Soluble in hydrofluoric acid. Insoluble in water. | |
[Si]=[Ti]=[Si] | |
104.037 | |
104.07 | |
(metals basis) |
Safety and Handling
GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H228-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 234-904-3
missing translation for 'rtecsNumber' : XR2420000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only