Learn More
Titanium silicide, 99.5% (metals basis), Thermo Scientific Chemicals
130.00 € - 538.00 €
Chemical Identifiers
CAS | 12067-57-1 |
---|---|
Molecular Formula | Si3Ti5 |
Molecular Weight (g/mol) | 323.59 |
MDL Number | MFCD01310208 |
InChI Key | GNKTZDSRQHMHLZ-UHFFFAOYSA-N |
Synonym | silicon; titanium, titanium silicide ti5si3, pentatitanium trisilicide, titanium silicide,-325 mesh |
PubChem CID | 6336553 |
IUPAC Name | silicon;titanium |
SMILES | [Si].[Si].[Si].[Ti].[Ti].[Ti].[Ti].[Ti] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11362178
|
Thermo Scientific Alfa Aesar
013087.18 |
50 g |
130.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11372178
|
Thermo Scientific Alfa Aesar
013087.30 |
250 g |
538.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as source, gate and drain contacts and local interconnections in CMOS integrated circuits. it is used in the C54 phase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12067-57-1 | |
323.59 | |
GNKTZDSRQHMHLZ-UHFFFAOYSA-N | |
6336553 | |
[Si].[Si].[Si].[Ti].[Ti].[Ti].[Ti].[Ti] |
Si3Ti5 | |
MFCD01310208 | |
silicon; titanium, titanium silicide ti5si3, pentatitanium trisilicide, titanium silicide,-325 mesh | |
silicon;titanium |
Specifications
2130°C | |
Powder | |
Si3Ti5 | |
silicon; titanium, titanium silicide ti5si3, pentatitanium trisilicide, titanium silicide,-325 mesh | |
GNKTZDSRQHMHLZ-UHFFFAOYSA-N | |
silicon;titanium | |
6336553 | |
Titanium Silicide |
12067-57-1 | |
99.5%, metal basis | |
MFCD01310208 | |
Soluble in hydrofluoric acid. Insoluble in water. | |
[Si].[Si].[Si].[Ti].[Ti].[Ti].[Ti].[Ti] | |
323.59 | |
323.76 |
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. This material and/or its container must be disposed of as hazardous waste.
P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H228-H315-H319-H335
missing translation for 'dotInformation' : DOT Class: 4.1; Packaging Group: III
missing translation for 'einecsNumber' : 235-082-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only