missing translation for 'onlineSavingsMsg'
Learn More

Titanium silicide sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.5% (metals basis), Thermo Scientific™

Product Code. 15463815
Click to view available options
Quantity:
1 Ea.
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Ea.
MDL Number MFCD00151541
Formula Weight 104.07
Percent Purity 99.5%
Assay (metals basis)
Chemical Name or Material Titanium silicide sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'