missing translation for 'onlineSavingsMsg'
Learn More

Titanium sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.7% (metals basis), Thermo Scientific Chemicals

Product Code. 15494825
Click to view available options
Quantity:
1 Ea.
This item is not returnable. View return policy
This item is not returnable. View return policy

 

Titanium sputtering target and its alloys are characterized by their lightness, strength and corrosion resistance and are used widely in aerospace applications. In addition, these properties also make the material suitable for medical applications (e.g. replacement hip joints). Titanium dioxide, TiO2 is used as a white pigment in paints and plastics as it provides great opacity. The same material is also used in the manufacture of heat resisting and durable glass, the TiO2 replacing certain proportions of the soda. Titanium carbide is used to manufacture cemented carbides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Ea.
CAS 7440-32-6
Odor Odorless
Molecular Formula Ti
MDL Number MFCD00011264,MFCD00151301
Synonym oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
Solubility Information Insoluble in water.
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
SMILES [Ti]
IUPAC Name titanium
Molecular Weight (g/mol) 47.87
PubChem CID 23963
ChEBI CHEBI:33341
Percent Purity 99.7%
Assay (metals basis)
Chemical Name or Material Titanium sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'