missing translation for 'onlineSavingsMsg'
Learn More
Titanium Standard Metal Solution 1000ppm, for AAS, Solution HCl 2M, Fisher Chemical™ Product Code.: 10611193

Titanium Standard Metal Solution 1000ppm, for AAS, Solution HCl 2M, Fisher Chemical™

Product Code. 10611193
100 mL
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy

Product Code. 10611193

Brand: Fisher Chemical J/8110/05

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Titanium Standard Metal Solution 1000ppm, for AAS, Solution HCl 2M

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number 11264

Specifications

Packaging HDPE plastic bottle
Quantity 100 mL
Physical Form Liquid
Color Purple
compliance-icons
Product Identifier
  • Titanium solution 1000 ppm
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Titanium Standard Metal Solution 1000ppm, for AAS, Solution HCl 2M, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.