missing translation for 'onlineSavingsMsg'
Learn More
Titanium Thinfoil, 0.0114mm (0.00045in) thick, 99.6% (metals basis), Thermo Scientific Chemicals Product Code.: 11359768

Titanium Thinfoil, 0.0114mm (0.00045in) thick, 99.6% (metals basis), Thermo Scientific Chemicals

Product Code. 11359768
200 x 200 mm
Click to view available options
Quantity:
100 x 100 mm
200 x 200 mm
This item is not returnable. View return policy

Product Code. 11359768

Brand: Thermo Scientific Alfa Aesar 044242.GW

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-32-6
Molecular Formula Ti
Molecular Weight (g/mol) 47.87
MDL Number MFCD00011264,MFCD00151301
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
Synonym oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
PubChem CID 23963
ChEBI CHEBI:33341
IUPAC Name titanium
SMILES [Ti]

Specifications

Quantity 200 x 200 mm
Percent Purity 99.6%
Grade Commercial Pure 4
Odor Odorless
Weight ≈0.514g/100x100mm
Form Thin Foil
Assay (metals basis)
Chemical Name or Material Titanium Thinfoil, 0.0114mm (0.00045 in.) thick, Light Tight

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT17K026 Certificate TypeCertificate of Analysis Date26/07/2024 Catalog Number044242.GW,044242.GH
Lot NumberS26K036 Certificate TypeCertificate of Analysis Date02/07/2024 Catalog Number044242.GW,044242.GH
Lot NumberQ26I056 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog Number044242.GW,044242.GH
Lot NumberW12F019 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number044242.GW,044242.GH
Lot NumberW30B015 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number044242.GW,044242.GH

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Titanium Thinfoil, 0.0114mm (0.00045in) thick, 99.6% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.