missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium wire, 0.75mm (0.03in) dia, 99.98% (metals basis), Thermo Scientific Chemicals
53.57 € - 144.72 €
Chemical Identifiers
CAS | 7440-32-6 |
---|---|
Molecular Formula | Ti |
Molecular Weight (g/mol) | 47.87 |
MDL Number | MFCD00011264,MFCD00151301 |
InChI Key | RTAQQCXQSZGOHL-UHFFFAOYSA-N |
Synonym | oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1 |
PubChem CID | 23963 |
ChEBI | CHEBI:33341 |
IUPAC Name | titanium |
SMILES | [Ti] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11376537
|
Thermo Scientific Alfa Aesar
000361.BU |
25 cm |
53.57 €
25cm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11366537
|
Thermo Scientific Alfa Aesar
000361.BY |
100 cm |
144.72 €
100cm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-32-6 | |
47.87 | |
RTAQQCXQSZGOHL-UHFFFAOYSA-N | |
23963 | |
titanium |
Ti | |
MFCD00011264,MFCD00151301 | |
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1 | |
CHEBI:33341 | |
[Ti] |
Specifications
7440-32-6 | |
MFCD00011264,MFCD00151301 | |
RTAQQCXQSZGOHL-UHFFFAOYSA-N | |
titanium | |
23963 | |
99.98% | |
≈1.99g/m | |
(metals basis) |
Ti | |
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1 | |
[Ti] | |
47.87 | |
CHEBI:33341 | |
Odorless | |
Wire | |
Titanium wire, 0.75mm (0.03 in.) dia., Temper: as drawn |
Safety and Handling
missing translation for 'einecsNumber' : 231-142-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only