missing translation for 'onlineSavingsMsg'
Learn More

SI Analytics TitroLine™ 5000 with 20 ml Basic Dosing Unit

TitroLine™ 5000 with 20 ml Dosing Basic Unit with Ag-electrode ideal for use in many laboratory and scientific and research environments.

Brand:  SI Analytics 285225850

3313.56 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.


Product Code. 15334408

  • 3800.00 € / Each
Estimated Shipment: 18-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • TitroLine™ 5000 with 20 ml Dosing Basic Unit with Ag-electrode
  • Automatic Titrators TitroLine™ 5000
  • Very compact new titrator for simple routine titrations
  • Titrations to pH, mV and μA endpoint (2 EP)
  • Titrations to one equivalence point (pH and mV) with dynamic or linear titration addition
Typical applications for TitroLine™ 5000:
  • Salt content in foodstuffs (cheese, soy sauce, ketchup)
  • Total acidity in wine and beverages
  • Nitrogen according to Kjeldahl
  • Chloride in drinking water and other samples
  • Alkalinity (p + m-value)
  • FOS/TAC
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TitroLine™ 5000 benchtop titrator
20 ml Basic Dosing Unit
TitroLine™ 5000
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.