missing translation for 'onlineSavingsMsg'
Learn More

SI Analytics™ TitroLine™ 5000 Automatic Titrator

Product Code. p-7070894 Shop All SI Analytics Products
Click to view available options
Model:
Titrator, 20 mL Dosing Unit
Titrator, 50 mL Dosing Unit
Type:
Automatic Titrator
Titrator with Electrode
Unit Size:
Each
This item is not returnable. View return policy

Artikelnummer. 15314408

Marke: SI Analytics™ 285225760

Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Beinhaltet

Cat. No. 15314408 - TitroLine 5000 with 20mL dosing unit
Ready to use assembled 20mL dosing unit, manual controller, titration clamp, stand rod, magnetic stirrer TM 50 and power supply 100 to 240V

Cat. No. 15324408 - TitroLine 5000 with 50mL dosing unit
pH electrode and buffer set, with ready to use assembled 50mL dosing unit, manual controller, titration clamp, stand rod, magnetic stirrer TM 50 and power supply 100 to 240V

Easily obtain precise and quick results without the need for training or knowledge of automatic titration. SI Analytics™ TitroLine™ 5000 Automatic Titrator is the first choice if you are looking for an easy to use automatic titrator for any application.

  • High resolution pH/mV-measurement input for pH-, ORP-, silver - and further mV-electrodes
  • pH/mV-input with electrode socket according to DIN 19 262 or additional with BNC socket insert (Z 860)
  • Pt 1000 and NTC 30 temperature measurement input (socket 2 x 4mm) for automatic temperature compensation
  • Pre-installed standard methods for FOS/TAC, alkalinity, total acidity in drinks, chloride
  • Unit operated using the keys on the device itself, the controller TZ 3880 and optional PC-keyboard (USB)
  • Graphics-capable TFT display
  • Linear and dynamic titration to equivalence points
  • Titration to pH and mV-ends points
  • Stirrer Connection: TM 50 power supply directly through piston burette
  • High resolution pH/mV-measurement input for pH-, ORP-, silver - and further mV-electrodes
  • Same manual titration and dosing function as the piston burette Titronic™ 300
  • Typical applications:
    • pH-value, alkalinity (p+m-value)
    • FOS/TAC
    • Total Kjeldahl nitrogen
    • Permanganate index and COD
    • Chloride in wastewater

Zertifizierungen

CE-mark: EMC: 2004/108/EG; safety EG- Directive 2006/95

Compliance

ISO 8655, part 6

TRUSTED_SUSTAINABILITY

Spezifikation

Certifications/Compliance ISO 8655, part 6, CE-mark: EMC: 2004/108/EG; safety EG- Directive 2006/95
Connector Types 1 x USB-A and 1 x USB-B, 2 x RS-232-C
Description Automatic Titrator
Display Type Graphics-capable TFT
Type Automatic Titrator
For Use With (Application) Determine chloride in wastewater, permanganate index and COD, total Kjeldhal nitrogen, FOS/TAC, pH value, alkalinity
Includes Basic unit without electrode, with ready to use assembled 20 ml dosing unit, manual controller, titration clamp, stand rod, magnetic stirrer TM 50 and power supply 100-240 V
Model Titrator, 50 mL Dosing Unit
Resolution Display resolution: 0.005 to 0.025mL; Burette resolution: 8000
Temperature Range (Metric) Ambient 10°C to 40°C
Width (Metric) 13.5 cm
Depth (Metric) 20.5 cm
Electrical Requirements 100 to 240V; 50/60Hz, power input 30 VA
Voltage 100/240 V
Frequency 50/60 Hz
Mehr anzeigen Weniger anzeigen
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt