Learn More
SI Analytics™ TitroLine™ 7000 Basic Unit Model TL 7000-M1
Features the highest safety for measuring and calibrating through wireless sensor recognition for ID-electrodes. SI Analytics™ TitroLine™ 7000 Basic Unit Model TL 7000-M1 possesses general titration feautres but offers improved performace.
Brand:  SI Analytics™ 285220120
Product Code. 15228954
  • 5705.00 € / Each
Estimated Shipment: 01-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • As a rule 10 - 15 user methods are usually enough for the most requirements. But sometimes you need a little bit more capacity. The TitroLine™ 7000 can store up to 50 user methods.
  • Highest safety for measuring and calibrating through wireless sensor recognition for TitroLine™ SI Analytics ID-electrodes.
  • The new electrodes with definite identification are sending their specific data wireless to the titrator. Therefore the TitroLine™ 7000 uses always the correct calibration data. Erroneous measurements are excluded.
  • With a pH stat application a given pH is first adjusted and then kept constant at the certain time with an acid or a base. The pH stat titration is applied to e.g.
    • the determination of the enzyme activity
    • the pH stat elution of soil sample at pH 4
    • the monitoring of the pH value during chemical syntheses
  • It is possible to detect and calculate up to 2 equivalence points during one titration. Thanks to this it is possible to determine also the Calcium and Magnesium hardness instead of only the total hardness.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TitroLine 7000 Basic Unit
Polypropylene
10°C to 40°C
15.3 cm
100 to 240v
Magnetic Stirrer TM235
TL 7000-M1
45 cm
29.6 cm