Learn More
SI Analytics TitroLine™ 7500 KF Trace M4
Fast, reliable, easy-to-use, and precise, the TitroLine™ 7500 KF trace is the specialist for low water contents.
Brand:  SI Analytics 285220890
Product Code. 15110226
  • 7830.00 € / Each
Estimated Shipment: 25-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • TitroLine™ 7500 KF trace M4
  • The TitroLine™ 7500 KF trace is the specialist for low water contents
  • Fast, easy and precise
  • With standard methods for different applications (titer determination, blank value…)
  • High visible full color display, that can be easily viewed from a distance and extreme angles
  • Storage of results via USB port (PDF and CSV -format) 
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Benchtop Titrator
Titrator unit, generator electrode TZ 1753 with diaphragm, TM 235 KF titration stand with integrated stirrer and pump, titration vessel TZ 1754, micro-dual platinum electrode KF 1150
Titrator unit, generator electrode TZ 1753 with diaphragm, TM 235 KF titration stand with integrated stirrer and pump, titration vessel TZ 1754, micro-dual platinum electrode KF 1150