missing translation for 'onlineSavingsMsg'
Learn More

SI Analytics™ TitroLine™ 7500 Volumetric Titrators

Easy and precise, with standard methods for different applications. SI Analytics™ TitroLine™ 7500 Volumetric Titrator offers more performance and is the volumetric generalist for a wide range of use.

Brand:  SI Analytics™ 285220810

 View more versions of this product

Product Code. 15157982

  • 6925.00 € / Each
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Fast, easy and precise
  • With standard methods for different applications (titer determination, blank value…)
  • High visible full color display, that can be easily viewed from a distance and extreme angles
  • Storage of results via USB port (PDF and CSV -format)
  • With intelligent interchangeable modules (only TitroLine™ 7500 KF)
  • Applications for the TitroLine 7500™ KF: KF volumetry, dead-stop-titrations (SO2, bromine number)
 
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DIN EN ISO 8655
KF volumetry, dead-stop-titrations (SO2,bromine number).
Polypropylene
10°C to 40°C
15.3 cm
100 to 240v
Volumetric KF-Titrator
Interchangeable Module WA 05
TL 7500 KF 05
45 cm
29.6 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers